AGT™ Dry Media Format Procurement Benefits
1:01
"Discover the QC-friendly media format with fewer steps and a faster workflow: AGT Dry Media Format from Thermo Fisher. Save time, resources and money. Benefits include mimized risks and consistent scale-up from research to production. AGT media are manufactured using granulation technology. We blend multiple dry and liquid components into a lattice-like granular structure. It’s a complete dry media format for your large-scale manufacturing process. Reduce your total cycle costs by decreasing time involved in raw material planning, procurement and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media. For more information visit https://www.thermofisher.com/agt AGT technology combines complex media components into a single-part, homogeneous formulation, which is automatically adjusted for pH and osmolality, is highly concentrated, and is QC-friendly due to fewer steps and a faster workflow, helping you save valuable time, resources, and money to allow for minimal risk and consistent scale-up from research to production. AGT media are manufactured using granulation technology that blends multiple dry and liquid components into a lattice-like, granular structure, which enables rapid rehydration and dissolution from a singular, dry format. AGT dry media format from Gibco. A complete dry media format for your large-scale manufacturing process. Learn more at https://www.thermofisher.com/us/en/home/life-science/bioproduction/gibco-bioprocessing/media-formats/agt.html