AGT Dry Media Format R&D Benefits | Gibco
0:52
Description
Related Videos
AGT is a dry granular media format that enables rapid rehydration and dissolution. AGT is a complete medium that is pH and osmolality pre-adjusted. It offers all the benefits of liquid media without the cost, storage, and transportation issues. You will benefit from high consistency and productivity across all stages of production: from development to commercial manufacturing. Learn more at https://www.thermofisher.com/us/en/home/life-science/bioproduction/gibco-bioprocessing/media-formats/agt.html