Back to search results for "Differentiating neurons"
Stem cell research-How to use Gibco® PSC Neural Induction Medium
3:20
Switching the growth medium to Gibco Neural Induction media can reach confluence within seven days. You’ll change media every other day. Colonies with a uniform morphology indicate a good neural induction. Induced NSCs can then be disassociated and expanded in neural expansion medium. Expanded NSCs can be stained with antibodies against neural markers such as Sox1, Nestin and Sox2. Expanded NSCs can also be differentiated into specific neural cell types, such as neurons, astrocytes and oligodendrocytes. Learn more at https://www.thermofisher.com/order/catalog/product/A1647801