How to Measure PCR Efficiency of an Assay - Ask TaqMan Episode 4
3:21
Description
Related Videos
To check the efficiency of an assay, you need to run a template dilution curve in real-time, and then look at the resulting slope of that curve.