      • Stem Cell Research
      Applied Biosystems™ VetMAX™ PRRSV EU & NA 2.0 Kit
      1:08
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The Applied Biosystems™ VetMAX™ PRRSV Europe and North America 2.0 Kit has the most documented detection of variants of any kit on the market. Internal and external validation studies done on more than 780 field samples of 13 origins have proven the kit’s performance on all four subtypes of the EU PRRS virus. This ready-to-use triplex kit uses an exogenous internal positive control, fast cycling, and simple protocols to deliver results in under two hours. With best-in-class support and continuous global monitoring, you can be sure you have the most up-to-date design.
      View More
      View Less
      Share
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels