      • Stem Cell Research
      Invitrogen Bigfoot Cell Sorter - Biosafety
      1:47
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      An integrated custom-designed biosafety cabinet ensures the safety of users and samples and is ergonomically optimized for sorting workflows. The system has been verified to be functionally equivalent to the personnel and product protection standards for a Class II Type A2 biosafety cabinet per NSF International Standard 49. The adjustable sash maintains containment while allowing easy access to the nozzle and sample/sort areas, providing constant user protection.
      View More
      View Less
      Learn more about the Invitrogen Bigfoot Cell Sorter
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels