      • Stem Cell Research
      Invitrogen Bigfoot Cell Sorter - Time Savings and Higher Throughput
      1:49
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The Bigfoot Spectral Cell Sorter offers sort rates of >70,000 eps and analysis rates of >100,000 eps. The system is also capable of multi-way sorting into 1.5ml, 5ml, 15ml, and 50ml tubes with an easily configurable sort output tube holder. Using tubes, the sorter is capable of 6-way sorting and virtual 18-way sorting. Bigfoot can also perform 4-way sorting into 96 well plates, 8-way sorting into 384 well plates, and straight down sorting into 1536 well plates. The speed and recovery of the Bigfoot Spectral Cell Sorter is designed to exceed your expectations.
      View More
      View Less
      Learn more about the Invitrogen Bigfoot Cell Sorter
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels