      • Stem Cell Research
      Affinity Chromatography: Innovative Approach for Enhanced Yield and Purity in Plasma-derived IgG
      31:23
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Traditional methods for producing Intravenous Immunoglobulin (IVIG) have been relying on ethanol precipitation cascades, often demanding compromises in yield and purity. We present a versatile approach utilizing direct adsorption of plasma IgG as well as recovery of IgG from side fractions via affinity chromatography. This innovative technique significantly enhances IgG yield while fulfilling critical quality attributes for therapeutic IVIG. IgG subclass distribution is maintained identical to that found in plasma and the resulting IgG is fully functional and comparable to commercial IVIG preparations, as demonstrated by Fc in-vitro assays. Our method achieves a substantial reduction of impurities such as IgA, IgM, and FXI/FXIa in a single step, thereby streamlining the purification process and improving product quality. Furthermore, the affinity chromatography resin employed in this process can be re-used multiple times, enhancing the cost-effectiveness and sustainability of the method. By eliminating the need for complex and time-consuming ethanol precipitation cascades, our method offers a more efficient and cost-effective alternative for IVIG production. In addition, this technique can further increase yield by affinity capture of IgG from side fractions where it is traditionally lost. This presentation will show details of the process performance, comparative analyses, and the significant advantages of this novel approach.
      View More
      View Less
      Learn more about Purification Plasma Fractionation
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels