      • Stem Cell Research
      Gibco CHOvantage GS Cell Line Development Kit
      1:27
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      For cell line development scientists aiming to achieve stable, high-titer expression and consistent protein quality efficiently, Gibco™ CHOvantage™ GS offers an innovative and cost-effective solution. Designed as a coordinated workflow, the platform enables you to move from transfection through clone selection using compatible components intended to support bioprocess development and manufacturing applications. An optimized media and feed system (Gibco Efficient Pro Medium (+) Insulin, Feed 3, and Feed Enhancer) are included to support robust cell growth and high protein expression ≥7 g/L for mAbs. Unlike legacy vector platforms, CHOvantage leverages a transposase-based system to reduce clone screening time and help ensure consistent protein quality. CHOvantage facilitates production of highly productive pools within 4 weeks and clones in as little as 14 weeks. Royalty-free commercial licensing is available to help significantly reduce development costs.
      View More
      View Less
      Learn more about the Gibco CHOvantage GS cell line development kit.
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels