      • Stem Cell Research
      Fast, Flexible, and Spectrally Brilliant: Invitrogen Attune Xenith Flow Cytometer
      1:00
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis with the Invitrogen Attune Xenith Flow Cytometer, powered by our legacy core acoustic hydrodynamic focusing technology. Experience remarkable speed, operating five times faster than most spectral cytometers, and benefit from innovative flexibility with seamless transitions between spectral unmixing and conventional flow cytometry on the same instrument using the same sample. Designed to handle challenging samples without clogging, the Attune Xenith Flow Cytometer boasts an advanced optical design featuring 6 lasers, 51 fluorescence detectors, and 6 scatter channels. Enjoy simplified maintenance with an intuitive touchscreen and advanced remote access diagnostics, including onboard cameras to ensure uninterrupted performance. Learn how the Invitrogen Attune Xenith Flow Cytometer can transform your research with speed, flexibility, and unparalleled performance. Visit thermofisher.com/attunexenith for more information.
      View More
      View Less
      Watch now
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Recently Published Videos
      • Play video How to choose: Thermo Scientific™ lab refrigerator and freezer accessories
        How to choose: Thermo Scientific™ lab refrigerator and freezer accessories
        Options and accessories to organize your cold storage including stainless steel drawers, sliding basket drawers, chromatography shelf support, enzyme bins
        2:15
      • Play video HPLC and UHPLC column selection guide
        HPLC and UHPLC column selection guide
        Take a quick tour of the selection guide and see how easy it is to find the right HPLC and UHPLC column for your application.
        0:26
      • Play video PowerFlex Thermal Cycler​ Cinematic
        PowerFlex Thermal Cycler​ Cinematic
        PowerFlex Thermal Cycler delivers precise PCR optimization, broad format compatibility (96, 3×32, 2x384), and flexible throughput—see it in action!
        1:39
      • Play video Introducing: Thermo Scientific™ TSC Series Recirculating Chillers
        Introducing: Thermo Scientific™ TSC Series Recirculating Chillers
        See why Thermo Scientific™ TSC Series Recirculating Chillers are the reliable choice for sustainability, versatility, and precise temperature control.
        1:13
      • Play video Toward liquid biopsy in kidney transplantation
        Toward liquid biopsy in kidney transplantation
        Join Jamil R. Azzi, MD, PhD, as he explores emerging approaches to kidney transplant technology, health, and patient care.
        29:14
      • Play video Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        In this episode of Absolute Gene-ius, Lexi Heger, PhD candidate at Michigan State University, explores how qPCR and digital PCR are transforming plant pathology.
        36:05
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels