Spatial Biology Antibodies: General consideration and validation
43:31
Description
Related Videos
Webinar recording which discusses the importance of choosing conjugated primary antibodies for multiplexing in tissue samples to increase sensitivity, reduce spectral overlap and reduce background fluorescence.