      • Stem Cell Research
      Affinity Chromatography: Innovative Approach for Enhanced Yield and Purity in Plasma-derived IgG
      31:23
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Traditional methods for producing Intravenous Immunoglobulin (IVIG) have been relying on ethanol precipitation cascades, often demanding compromises in yield and purity. We present a versatile approach utilizing direct adsorption of plasma IgG as well as recovery of IgG from side fractions via affinity chromatography. This innovative technique significantly enhances IgG yield while fulfilling critical quality attributes for therapeutic IVIG. IgG subclass distribution is maintained identical to that found in plasma and the resulting IgG is fully functional and comparable to commercial IVIG preparations, as demonstrated by Fc in-vitro assays. Our method achieves a substantial reduction of impurities such as IgA, IgM, and FXI/FXIa in a single step, thereby streamlining the purification process and improving product quality. Furthermore, the affinity chromatography resin employed in this process can be re-used multiple times, enhancing the cost-effectiveness and sustainability of the method. By eliminating the need for complex and time-consuming ethanol precipitation cascades, our method offers a more efficient and cost-effective alternative for IVIG production. In addition, this technique can further increase yield by affinity capture of IgG from side fractions where it is traditionally lost. This presentation will show details of the process performance, comparative analyses, and the significant advantages of this novel approach.
      View More
      View Less
      Learn more about Purification Plasma Fractionation
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Recently Published Videos
      • Play video Gibco CHOvantage GS Cell Line Development Kit
        Gibco CHOvantage GS Cell Line Development Kit
        Advance from transfection to high-producing CHO clones faster with the Gibco CHOvantage cell line development workflow. Achieve strong productivity, stable expression, consistent quality and scalable performance to support biotherapeutic development.
        1:27
      • Play video Introducing: Thermo Scientificâ„¢ TSC Series Recirculating Chillers
        Introducing: Thermo Scientificâ„¢ TSC Series Recirculating Chillers
        See why Thermo Scientificâ„¢ TSC Series Recirculating Chillers are the reliable choice for sustainability, versatility, and precise temperature control.
        1:13
      • Play video HPLC and UHPLC column selection guide
        HPLC and UHPLC column selection guide
        Take a quick tour of the selection guide and see how easy it is to find the right HPLC and UHPLC column for your application.
        0:26
      • Play video How emerging technologies are filling the gaps and positively impacting post-transplant care
        How emerging technologies are filling the gaps and positively impacting post-transplant care
        Multi-organ transplant recipient and patient advocate Valen Keefer shares her perspective on solutions shaping transplant patients' experiences.
        4:41
      • Play video Toward liquid biopsy in kidney transplantation
        Toward liquid biopsy in kidney transplantation
        Join Jamil R. Azzi, MD, PhD, as he explores emerging approaches to kidney transplant technology, health, and patient care.
        29:14
      • Play video How to operate: Thermo Scientific Multidrop Pico 8 sf Creating protocols and editing fluid parameters
        How to operate: Thermo Scientific Multidrop Pico 8 sf Creating protocols and editing fluid parameters
        Learn how to create a protocol and edit fluid parameters for the Thermo Scientificâ„¢ Multidropâ„¢ Pico 8 sf Digital Dispenser using Multidrop PicoIT software.
        2:28
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels