      • Stem Cell Research
      CHO Cell Line Development with CHOvantage GS Kit
      1:27
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Develop high-titer CHO cell lines faster with the CHOvantage GS Cell Line Development Kit. Achieve >7 g/L titers, stable clones in 14 weeks, and scalable workflows using transposon-based technology for consistent biologics production. Achieving high titers while maintaining consistency and scalability is one of the biggest challenges in CHO cell line development. The Gibco CHOvantage GS Cell Line Development Kit is designed to accelerate timelines while improving performance and reproducibility. Key capabilities include: 1) Achieve >7 g/L monoclonal antibody titers 2) Generate stable clones in as little as 14 weeks 3) Develop highly productive pools in 4 weeks 4) Utilize a transposon-based vector system for consistent gene integration 5) Reduce clone-to-clone variability and improve predictability 6) Support scalable production with stability up to 100 generations 7) Enable efficient transition from development to manufacturing The CHOvantage GS system integrates optimized workflows, advanced vector design, and Gibco media systems to help accelerate biologics development while maintaining quality and regulatory readiness.
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Recently Published Videos
      • Play video Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Fluid Path
        Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Fluid Path
        This video showcases a fluid path for the CTS Compleo System for formulation and controlled filling into output containers for cell therapy manufacturing.
        2:47
      • Play video How to operate: Thermo Scientific Nalgene and Thermo Scientific Nunc color-capped sample storage tubes
        How to operate: Thermo Scientific Nalgene and Thermo Scientific Nunc color-capped sample storage tubes
        Organize samples, projects, and experiments using color caps and inserts for your storage tubes to improve clarity, accuracy, and workflow management.
        2:06
      • Play video Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Sampling
        Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Sampling
        This video showcases one method of aseptic sampling performed using the Gibco CTS Compleo Fill and Finish System for cell therapy manufacturing.
        1:25
      • Play video Single-use bioreactor scaling calculator demo
        Single-use bioreactor scaling calculator demo
        Learn how to use a single-use bioreactor scale-up calculator to simplify complex bioprocess calculations, improve accuracy, and support efficient development and production workflows.
        8:35
      • Play video Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool, your key to finding the proper replacement for your current cell culture reagent supplier.​
        0:28
      • Play video Aegis Film for DynaDrive Single-Use Bioreactors
        Aegis Film for DynaDrive Single-Use Bioreactors
        Pair DynaDrive single-use bioreactors with Aegis film to simplify validation and support global compliance. PFAS- and Ergifos-168–free, it delivers strength and compatibility from media prep through production.
        1:02
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels