      • Stem Cell Research
      Gibco CHOvantage GS Cell Line Development Kit
      1:27
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      For cell line development scientists aiming to achieve stable, high-titer expression and consistent protein quality efficiently, Gibco™ CHOvantage™ GS offers an innovative and cost-effective solution. Designed as a coordinated workflow, the platform enables you to move from transfection through clone selection using compatible components intended to support bioprocess development and manufacturing applications. An optimized media and feed system (Gibco Efficient Pro Medium (+) Insulin, Feed 3, and Feed Enhancer) are included to support robust cell growth and high protein expression ≥7 g/L for mAbs. Unlike legacy vector platforms, CHOvantage leverages a transposase-based system to reduce clone screening time and help ensure consistent protein quality. CHOvantage facilitates production of highly productive pools within 4 weeks and clones in as little as 14 weeks. Royalty-free commercial licensing is available to help significantly reduce development costs.
      View More
      View Less
      Learn more about the Gibco CHOvantage GS cell line development kit.
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Recently Published Videos
      • Play video Cell Culture Confessions: The Strange, Sticky, and Surprisingly Common Lab Questions
        Cell Culture Confessions: The Strange, Sticky, and Surprisingly Common Lab Questions
        From sticky cells to floating mysteries, every scientist’s got a story. Join Kevin and Sara for real lab confessions, laughs, and culture-saving tips.
        17:43
      • Play video HPLC and UHPLC column selection guide
        HPLC and UHPLC column selection guide
        Take a quick tour of the selection guide and see how easy it is to find the right HPLC and UHPLC column for your application.
        0:26
      • Play video Toward liquid biopsy in kidney transplantation
        Toward liquid biopsy in kidney transplantation
        Join Jamil R. Azzi, MD, PhD, as he explores emerging approaches to kidney transplant technology, health, and patient care.
        29:14
      • Play video Aegis Film for DynaDrive Single-Use Bioreactors
        Aegis Film for DynaDrive Single-Use Bioreactors
        Pair DynaDrive single-use bioreactors with Aegis film to simplify validation and support global compliance. PFAS- and Ergifos-168–free, it delivers strength and compatibility from media prep through production.
        1:02
      • Play video Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool, your key to finding the proper replacement for your current cell culture reagent supplier.​
        0:28
      • Play video Clinical utility of MMDx: Diagnosing early antibody mediated rejection
        Clinical utility of MMDx: Diagnosing early antibody mediated rejection
        Join Rajeev Sharma, MD, FACS, as he explores two cases of antibody mediated rejection diagnosed with the help of MMDx.
        16:42
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels