Process 11 Parallel Twin-Screw Extruder - Overview
5:34
Starting with as little as 20 g/h of material, the Thermo Scientific Process 11 Twin-screw Extruder is designed to meet the key challenges faced by research and development formulation scientists. Complete compounding line including feeding solutions and downstream equipment such as water baths, conveyor belt, variable length pelletizers, melt pump, 3D filament spooler or sheet take off are available.