imPULSE Single-Use Mixer (S.U.M.) with Touchscreen
6:44
The Thermo Scientific imPULSE Single-Use Mixer (S.U.M) is ideal for a wide range of liquid-to-liquid or liquid-to-powder mixing applications. It features innovative mixing disc technology, an improved user experience with a Touchscreen Console, and is scalable from 30 liters up to 5,000 liters.