How to use the Invitrogen QuantiGene Plex Data Analysis App on the Thermo Fisher Connect Platform
12:35
"Learn how to use the Invitrogen™ QuantiGene Plex™ Analysis App available on Thermo Fisher Connect to analyze Invitrogen™ QuantiGene Plex™ assay results. In this video we will walk you step by step through the process of accessing and analyzing your Invitrogen™ QuantiGene Plex™ assay results. This video discusses the following steps to help you with your analysis: 1) Overview of the QuantiGene Plex data analysis workflow 2) How to import .csv output data files from the xPonent software and set up the plate map 3) Select data normalization genes 4) Review and edit QC criteria 5) Run data analysis 6) Export analyzed data as .csv files or generate .chp files for advance data analysis using TAC software 7) Advanced data analysis using Applied Biosystems™ Transcriptome Analysis Console (TAC) software For more information, please refer to the Invitrogen™ QuantiGene Plex™ user guide or contact Technical Support. Invitrogen™ QuantiGene Plex™ Assays: https://www.thermofisher.com/qgp Invitrogen™ QuantiGene Plex™ Analysis App: https://apps.thermofisher.com/apps/quantigene/ If you have any questions or concerns about running Invitrogen™ QuantiGene™ assay, please contact Thermo Fisher Scientific Technical Service: eutotech@thermofisher.com (Europe) or techservices@thermofisher.com (North America). Or visit our Support Centers: https://www.thermofisher.com/antibodysupport"