Applied Biosystems™ VetMAX™ PRRSV EU & NA 2.0 Kit
1:08
The Applied Biosystems™ VetMAX™ PRRSV Europe and North America 2.0 Kit has the most documented detection of variants of any kit on the market. Internal and external validation studies done on more than 780 field samples of 13 origins have proven the kit’s performance on all four subtypes of the EU PRRS virus. This ready-to-use triplex kit uses an exogenous internal positive control, fast cycling, and simple protocols to deliver results in under two hours. With best-in-class support and continuous global monitoring, you can be sure you have the most up-to-date design.