Automated Image Analysis with the Attune CytPix flow cytometer
1:15
The Attune CytPix flow cytometer combines acoustic focusing with a high-speed brightfield camera for simultaneous high throughput flow cytometry and high resolution brightfield imaging. Automated image analysis translates event features into distinct extended parameters that can be combined with standard flow parameters.