      • Stem Cell Research
      Industrial gas analysis by new OE-FTIR technology
      2:35
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Introducing the MAX-iR FTIR Gas Analyzer with new, optically enhanced FTIR (OE-FTIR) technology for fast, sensitive gas analysis in industrial environments. By providing real-time analysis and measurements down to single-digit parts-per-billion, the OE-FTIR industrial gas analyzer offers a more efficient alternative to traditional GC-based gas analysis systems. The MAX-iR analyzer forms the heart of several multi-channel systems built for ambient air monitoring, continuous emissions monitoring (CEM), ultra-high purity gas analysis, carbonation stream analysis and other industrial processes.
      View More
      View Less
      Learn more about the MAX-iR systems and optically enhanced FTIR (OE-FTIR) technology
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In What's new
      • Play video Gibco Efficient Pro Feed 3 Protein Production Workflow
        Gibco Efficient Pro Feed 3 Protein Production Workflow
        Boost CHO-K1 GS titers by up to 146% with Efficient-Pro Feed 3 and Feed Enhancer—a two-part, chemically defined, animal-origin-free system designed with advanced metabolomics to enhance productivity and maintain critical product quality.
        1:41
      • Play video Imaging made easy - EVOS Tip #40
        Imaging made easy - EVOS Tip #40
        Discover practical tips to help eliminate the complexities of microscopy without compromising performance - enabled by Invitrogen EVOS Imaging Systems.
        0:22
      • Play video PowerFlex Thermal Cycler​ Cinematic
        PowerFlex Thermal Cycler​ Cinematic
        PowerFlex Thermal Cycler delivers precise PCR optimization, broad format compatibility (96, 3×32, dual 384), and flexible throughput—see it in action!​
        1:39
      • Play video Imaging made easy - EVOS Tip #33
        Imaging made easy - EVOS Tip #33
        Discover practical tips to help eliminate the complexities of microscopy without compromising performance - enabled by Invitrogen EVOS Imaging Systems.
        0:22
      • Play video The Upstream Advantage: Unlocking bioprocessing potential
        The Upstream Advantage: Unlocking bioprocessing potential
        Hear from Thermo Fisher Scientific colleagues and learn about their perspectives on the future of bioprocessing.
        3:26
      • Play video Care & maintenance: Thermo Scientific TSX Series High-Performance Chromatography and TSG Series Sample Prep Refrigerators
        Care & maintenance: Thermo Scientific TSX Series High-Performance Chromatography and TSG Series Sample Prep Refrigerators
        Explore key safety features of Thermo Scientific TSX Series Chromatography and TSG Series Sample Prep refrigerators to help you keep them up and running.
        3:41
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels