      • Stem Cell Research
      Affinity Chromatography: Innovative Approach for Enhanced Yield and Purity in Plasma-derived IgG
      31:23
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Traditional methods for producing Intravenous Immunoglobulin (IVIG) have been relying on ethanol precipitation cascades, often demanding compromises in yield and purity. We present a versatile approach utilizing direct adsorption of plasma IgG as well as recovery of IgG from side fractions via affinity chromatography. This innovative technique significantly enhances IgG yield while fulfilling critical quality attributes for therapeutic IVIG. IgG subclass distribution is maintained identical to that found in plasma and the resulting IgG is fully functional and comparable to commercial IVIG preparations, as demonstrated by Fc in-vitro assays. Our method achieves a substantial reduction of impurities such as IgA, IgM, and FXI/FXIa in a single step, thereby streamlining the purification process and improving product quality. Furthermore, the affinity chromatography resin employed in this process can be re-used multiple times, enhancing the cost-effectiveness and sustainability of the method. By eliminating the need for complex and time-consuming ethanol precipitation cascades, our method offers a more efficient and cost-effective alternative for IVIG production. In addition, this technique can further increase yield by affinity capture of IgG from side fractions where it is traditionally lost. This presentation will show details of the process performance, comparative analyses, and the significant advantages of this novel approach.
      View More
      View Less
      Learn more about Purification Plasma Fractionation
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In What's new
      • Play video Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Fluid Path
        Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Fluid Path
        This video showcases a fluid path for the CTS Compleo System for formulation and controlled filling into output containers for cell therapy manufacturing.
        2:47
      • Play video Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool
        Try the Switch-to-Gibco Product Cross-Reference Tool, your key to finding the proper replacement for your current cell culture reagent supplier.​
        0:28
      • Play video Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Sampling
        Gibcoâ„¢ CTSâ„¢ Compleoâ„¢ Fill and Finish System Sampling
        This video showcases one method of aseptic sampling performed using the Gibco CTS Compleo Fill and Finish System for cell therapy manufacturing.
        1:25
      • Play video Single-use bioreactor scaling calculator demo
        Single-use bioreactor scaling calculator demo
        Learn how to use a single-use bioreactor scale-up calculator to simplify complex bioprocess calculations, improve accuracy, and support efficient development and production workflows.
        8:35
      • Play video Aegis Film for DynaDrive Single-Use Bioreactors
        Aegis Film for DynaDrive Single-Use Bioreactors
        Pair DynaDrive single-use bioreactors with Aegis film to simplify validation and support global compliance. PFAS- and Ergifos-168–free, it delivers strength and compatibility from media prep through production.
        1:02
      • Play video Absorbance, Redefined | Meet the New Multiskan Ease
        Absorbance, Redefined | Meet the New Multiskan Ease
        In 60 seconds, see a better way to run absorbance assays.
        0:55
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels