      • Stem Cell Research
      Back to search results for "Differentiating neurons"
      Stem cell research-How to use Gibco® PSC Neural Induction Medium
      3:20
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Switching the growth medium to Gibco Neural Induction media can reach confluence within seven days. You’ll change media every other day. Colonies with a uniform morphology indicate a good neural induction. Induced NSCs can then be disassociated and expanded in neural expansion medium. Expanded NSCs can be stained with antibodies against neural markers such as Sox1, Nestin and Sox2. Expanded NSCs can also be differentiated into specific neural cell types, such as neurons, astrocytes and oligodendrocytes. Learn more at https://www.thermofisher.com/order/catalog/product/A1647801
      View More
      View Less
      Learn more about PSC Neural Induction Medium
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      For "Differentiating neurons"
      • Play video Heart disease in a dish: Human iPSC-derived multicellular cardiac organoids
        Heart disease in a dish: Human iPSC-derived multicellular cardiac organoids
        This presentation showcases the development of an advanced 3D multicellular human heart tissue model for modelling cardiovascular diseases.
        51:39
      • Play video Personalized cell therapy for Parkinson’s disease
        Personalized cell therapy for Parkinson’s disease
        This presentation describes an established platform of core techniques for the production of mDA progenitors as a safe and effective therapeutic product.
        27:53
      • Play video Perfect Pairings: beta-3 Tubulin Monoclonal Antibody and ProLong RapidSet Antifade Mountant
        Perfect Pairings: beta-3 Tubulin Monoclonal Antibody and ProLong RapidSet Antifade Mountant
        Enable fast, reliable labeling and imaging of neuronal markers.
        0:17
      • Play video How to use Gibco PSC Neural Induction Medium
        How to use Gibco PSC Neural Induction Medium
        This video will show you how Gibco® PSC Neural Induction Medium generates up to 20 million neural stem cells from pluripotent stem cells in 7 days.
        3:14
      • Play video Training on Differentiating neurons
        Training on Differentiating neurons
        Jordan Dizon presents webinar on Differentiating neurons
        30:12
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels