imPULSE Single-Use Mixer (S.U.M.) with Touchscreen
6:44
Description
Related Videos
Watch this video to learn how the Thermo Scientific imPULSE Single-Use Mixer (S.U.M.) is an ideal mixing solution for liquid-to-liquid or liquid-to-powder mixing applications with capabilities from basic mixing to fully customized applications. Learn about the innovative, scalable mixing disc technology that is used in all vessels ranging from 30 L to 5,000 L. This mixing system also has a touchscreen console that provides superior integrated sensor monitoring, and pump control for automatic fill and harvest steps.