How to Use the Gibco CTS DynaCellect Bead Removal Kit (2:24)
2:25
Description
Related Videos
The Gibcoâ„¢ CTSâ„¢ DynaCellectâ„¢ Bead Removal Kit is specifically designed for depletion and bead removal from immune cells for cell processing applications. The bead removal kit consists of a 1L flow chamber, four PVC tubes, filters, and a cassette for easy installation.