How to optimize your parameters on the Neon NxT system
6:21
Description
Related Videos
This video shows you the key steps to optimize your parameters on the Neon NxT Electroporation System. By understanding the importance of factors like pulse voltage, pulse width, pulse number, cell density, cell health, and payload characteristics, you can achieve optimal performance, transfection efficiency, and cell viability. The video also highlights the significance of selecting the right electroporation buffer for specific applications. Don't miss this opportunity to learn how to maximize the potential of your Neon NxT system.