How to operate: Thermo Scientific ™ TSX™ Universal ULT Freezer What is universal voltage?
2:03
Description
Related Videos
The Thermo Scientific™ TSX™ Universal ultra-low temperature freezer is the next generation of the trusted TSX platform. The TSX Universal is truly a universal voltage freezer, capable of operating on global voltages between 100-230V in both 50/60Hz frequencies. In this video we will discuss universal voltage and how to power up the new TSX Universal ULT freezer.