      • Stem Cell Research
      SARS-CoV-2 pathogenesis and host response: Where are we in our understanding of the virus?
      32:25
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The ongoing SARS-CoV-2 pandemic has led to the loss of over 3M lives globally and economic loss of unparalleled scales. The virus belonging to the betacoronavirus genus shares its lineage with the 2003 SARS virus and can cause illness that ranges from mild respiratory tract infection to acute respiratory distress syndrome (ARDS) and death in severe cases. Distinct from its predecessors, several unique characteristics of SARS-CoV-2 has rendered this virus more infectious and altered its pathogenesis. For instance, the binding affinity of the receptor binding domain (RBD) region of the Spike protein to angiotensin converting enzyme 2 (ACE2), a key entry receptor is significantly higher than SARS (2003), a feature that contributes to the high transmissibility of the virus. Emerging variants of SARS-CoV-2 are an ongoing cause of concern as specific mutations in the RBD region have been linked to enhanced pathogenicity and virus fitness. Furthermore, the virus not only infects cells of the respiratory tract but also has been detected in several other vital organs including the heart and brain. Unsurprisingly, its wide tropism contributes to the diverse set of symptoms experienced by infected subjects. In addition to virus-related factors, the host response against the infection plays a significant role to determine the outcome of the disease. Factors such as genotypic background, inflammatory response as well as pre-existing comorbidities impact host susceptibility to SARS-CoV-2, course of infection and recovery time. Here, we will review our current understanding of SARS-CoV-2 pathogenesis through discussion of key discoveries that have advanced our knowledge on how this remarkable virus infects its host and causes disease.
      View More
      View Less
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Real-Time PCR Videos
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19EUA
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19EUA
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19quote
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19quote
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19CEIVD
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19CEIVD
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      • Play video Enhance your qPCR experience with innovative and intuitive features: Preview
        Enhance your qPCR experience with innovative and intuitive features: Preview
        Watch this short preview to learn about the innovative features offered on Applied Biosystemsâ„¢ QuantStudio RT-PCR Systems
        0:32
      • Play video Workflows for diagnostic testing and assay development with the QuantStudio 7 Pro Dx Real-Time PCR System
        Workflows for diagnostic testing and assay development with the QuantStudio 7 Pro Dx Real-Time PCR System
        The automated QuantStudioâ„¢ 7 Pro Dx Real-Time PCR System is an in vitro diagnostic device intended to perform fluorescence-based PCR to provide detection of nucleic acid sequences in human-derived specimens
        11:40
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels