      • Stem Cell Research
      How to prepare Essential 8 Medium for pluripotent stem cell culture
      1:32
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      This video will show you how to prepare complete Essential 8 Medium for the feeder-free culture of PSCs. Essential 8 Medium is a fully defined feeder-free medium, formulated for the growth and expansion of human pluripotent stem cells. It was originally developed in the laboratory of James Thomson, and validated by Cellular Dynamics International. To prepare 500 mL of complete Essential 8 Medium, aseptically mix the following components, 490 mL of Essential 8 Basal Medium, and 10 mL of Essential 8 Supplement. Complete Essential 8 Medium can be stored at 2-8°C for up to 2 weeks. Before using, warm the amount of complete medium required for that day at room temperature, until it is no longer cool to the touch. Don't warm the medium at 37°C. Given the lean nature of Essential 8 Medium, additional freeze-thawing of the supplement or complete medium is not recommended.
      View More
      View Less
      View our products
      Share
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels