      • Stem Cell Research
      How to adapt pluripotent stem cells from other PSC media into Essential 8 Medium
      4:09
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      This video will show you how to adapt PSCs adapted to feeder-based or feeder-free media to Essential 8 Medium and Vitronectin. [AUDIO TRANSCRIPT] This video will show you how to adapt PSCs adapted to feeder-based or feeder-free media to Essential 8 Medium and Vitronectin. Essential 8 Medium is a fully defined, feeder-free medium formulated for the growth and expansion of human pluripotent stem cells. It was originally developed as E8 Medium in the laboratory of James Thomson, and validated by Cellular Dynamics International. Unlike other feeder-free media such as mTeSR and StemPro hESC SFM, Essential 8 Medium does not require the presence of BSA (bovine serum albumin) that contributes to lot-to-lot variability. In addition, other serum-free media consist of more than 20 components, adding complexity, time, and cost, while Essential 8 Medium is comprised of only eight components. PSCs grown using mTeSR Medium on Matrigel-coated culture vessels can be thawed directly into Essential 8 Medium on vitronectin-coated culture vessels. PSCs maintained in KnockOut Serum Replacement on feeders prior to freezing should be thawed into KSR Medium on MEF feeder layer. Allow the cells to recover, and then passage into Essential 8 Medium on vitronectin-coated culture vessels, using EDTA as the passaging reagent. When thawing directly into Essential 8, place 10 mL of Essential 8 Medium in a 50-mL tube and warm to room temperature. Next, equilibrate an appropriate quantity of vitronectin-coated 6-well plates to room temperature. Remove the vial of PSCs from liquid nitrogen storage using metal forceps. Transfer it on dry ice to the cell culture hood. Immerse the vial in a 37°C water bath without submerging the cap. Swirl the vial gently. When only an ice crystal remains, remove the vial from the water bath. Spray the outside of the vial with 70% ethanol and place it in hood. Pipet cells gently into a sterile 50-mL conical tube using a 5-mL sterile pipette. Slowly add 10 mL of Essential 8 Medium drop-wise to cells in the 50-mL conical tube. While adding the medium, gently move the tube back and forth to mix the PSCs. This reduces osmotic shock to the cells. Rinse the vial with 1 mL of Essential 8 Medium and add to the 50-mL tube with cells. Slowly transfer the contents to a 15mL tube Centrifuge the cells at 200 g for 5 minutes. Aspirate and discard the supernatant. Resuspend the cell pellet in 2 mL of Essential 8 Medium by gently pipetting the cells up and down in the tube a few times. Aspirate the vitronectin from the pre-warmed vitronectin-coated plates. Slowly add the PSC suspension into the vitronectin-coated plate, plating one vial of thawed cells per well of the 6-well plate. Move the dish in several quick figure-eight motions to disperse cells across the surface of the wells. Place the dish gently into the 37°C, 5% CO2 incubator and incubate the cells overnight. The next day, replace the spent medium with fresh complete Essential 8 Medium. Replace the medium daily thereafter until the cells are approximately 85% confluent.
      View More
      View Less
      View our product
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels