      • Stem Cell Research
      MAGPIX System - How To Prepare Your Novex Assay
      2:08
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The move from multiple protein assays to multiplex assays is now totally within reach! The MAGPIX system is an affordable, compact, fluorescent-based detection system based on Luminex's proven xMAP technology. With the MAGPIX system every researcher can perform quantitative analysis of up to 50 proteins simultaneously using only 25μl of precious sample. The MAGPIX system is personal, benchtop instrument with out of the box set-up with step by step guide to interactive software and all designed for use with broad and expanding menu of assay kits. For a demonstration of the MAGPIX System, click here! One of the most important steps in running a successful multiplex assay on your MAGPIX system is making sure you prepare your sample correctly. To learn more about Novex singleplex and multiplex assay visit www.thermofisher.com/luminex. Just like any other experiment, you want to make sure you have cleared a section of your lab bench to work on and make sure you have everything you need on hand. Here's the list of all the items necessary for your multiplex assay. Make sure your MAGPIX System is routinely maintained, calibrated, and verified the day of use. It's also important that you make sure the samples that you'll be using are thawed immediately before your assay is set up. Remember that samples begin to degrade upon thawing, so the least amount of time your samples are at room temperature, the better. You'll need to clarify your samples by centrifugation at 14K for 10minutes. Samples can be stored on ice until they're loaded onto a plate. It's important to note that if you're using samples other than serum or plasma, you have 5mL of that media type available for standards that you'll use in an assay. Remove your Novex multiplex assay kit from the refrigerator at least one hour before you begin to allow equilibration to room temperature. While you're waiting, you can prepare the 1X wash solution by diluting 15mL of the 20X concentrate into 285mL of de-ionized water. In some cases, crystals may be present in the wash concentrate. If this occurs, warm the concentrate in a 37°C water bath until dissolved, or dilute the wash solution with crystals in 285mL of water and allow the wash solution to dissolve at room temperature. You'll want to prepare a wash station near a sink or dispensing reservoir for later use with your assay. It's also helpful to gather a small stack of paper towels that will be used for blotting your assay plate later on. You can now begin to set up the primary stage of your assay by following the steps located in the multiplex assay protocol supplied with the kit.
      View More
      View Less
      Learn more about MAGPIX System
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels