      • Stem Cell Research
      iBind Western System's Sequential Lateral Flow Technology Explained
      2:42
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Load and go- process western blots with sequential lateral flow technology using the Invitrogen iBind Western Device. How is it possible? The iBind Western device uses a technology called sequential lateral flow. Let’s examine how this technology works. A unique glass fiber matrix allows optimal and homogeneous flow of the detection solutions towards the stack region of the card. This technology allows one to use up to 80% less primary antibody and enables improved blot to blot consistency, after performing gel electrophoresis and transfer of the proteins to a solid support membrane such as PVDF or nitrocellulose. Proceed by rinsing the membrane in iBind solution and prepare the device for use. Once the iBind card is placed into the iBind device the membrane is placed onto the card after being immersed in iBind solution. Well one is loaded with primary antibody. Well 2 is loaded with iBind solution, then well 3 is loaded with the secondary antibody. Finally well 4 is loaded with the iBind solution. There is no power, pumps, vacuums or batteries required. Each of the well contents will move in sequential order across the membrane while the previous well contents are held in place until the previous well has completely drained. First the primary antibody flows across the membrane binding to protein. The second well will then flow across the membrane washing away the unbound primary antibodies. The third well with secondary antibody will flow across the membrane binding to remaining primary antibodies. The fourth and final well containing iBind solution flows across the membrane washing away unbound secondary antibodies. Once the final well has completely emptied and moved across the membrane you can remove and proceed to Western detection protocol. Sequential lateral flow technology. Powering every iBind western device from Thermo Fisher Scientific. Learn more at thermofisher.com/ibind
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels