      • Stem Cell Research
      Basics of PCR
      1:22
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Let’s now review the basic components of a PCR reaction. It starts with a strand of double-stranded template DNA that includes the target sequence. Also needed are primers—short, single-stranded DNA sequences that are complementary to a specific part of the target sequence. dNTPs, or the building blocks for new DNA strands, are included, as well as the DNA polymerase, the enzyme that uses single- stranded DNA as a template to synthesize a complementary DNA strand. The reaction runs in a buffer containing magnesium ions, which are crucial for the activity of the enzyme. While early PCR was performed by physically moving tubes from one water bath to another to incubate the reaction at the specific temperatures for each step of the process, today PCR is performed in a thermal cycler instrument, which automates the cycling of the reaction steps through the different temperatures. Thanks for watching!
      View More
      View Less
      Learn more about PCR
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels