      • Stem Cell Research
      Back to search results for "Differentiating neurons"
      How to use Gibco PSC Neural Induction Medium
      3:14
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      With Gibco PSC Neural Induction Medium, you can efficiently convert 1 million human pluripotent stem cells, or PSCs, into 20 million neural stem cells without laborious embryoid body formation or rosette isolation. Before neural induction, the quality of human PSCs is critical for highly efficient neural induction. High-quality PSCs with minimal or no differentiated colonies, as shown in the images, grown in feeder-free media, such as Essential 8 Medium or StemFlex Medium, are recommended. Any differentiated or partly differentiated colonies, as pointed with the arrow, should be removed before passaging. When PSCs reach 70% to 80% confluence,as shown in the image, passage and plate PSCs at a density of 25,000 to 30,000 cells/cm2 in a coated tissue culture dish. The use of 10 µM ROCK inhibitor during passaging has been shown to improve cell viability and attachment at this step. 24 hours after PSC splitting, the density of PSCs should be 15% to 25% confluence, as shown in the image. Simply switch the growth medium to Gibco PSC Neural Induction Medium to start neural induction. The total duration of neural induction is 7 days. Change Neural Induction Medium every other day during the 7 days of neural induction. During the 7 days, colonies with a uniform morphology indicate good neural induction. During neural induction, cells are keeping to proliferate, and cell density are increasing during induction process. As shown in the images, colonies with a non-neural morphology may be observed during neural induction due to differentiated PSCs being in the culture before neural induction. To remove them, mark the differentiated colonies with a microscopy marker and aspirate them with a Pasteur glass pipette when changing the medium. At day 7 of neural induction, cells reach confluence, as shown in the images. Induced NSCs can then be disassociated and expanded in neural expansion medium. Expanded NSCs can be stained with antibodies against neural markers, such as Sox1, nestin, and Sox2. Expanded NSCs can also be differentiated into specific neural cell types, such as neurons, astrocytes, and oligodendrocytes. Gibco Neural Induction Medium generates at least 20 million NSCs from 1 million PSCs in 7 days.
      View More
      View Less
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      For "Differentiating neurons"
      • Play video Personalized cell therapy for Parkinson’s disease
        Personalized cell therapy for Parkinson’s disease
        This presentation describes an established platform of core techniques for the production of mDA progenitors as a safe and effective therapeutic product.
        27:53
      • Play video Training on Differentiating neurons
        Training on Differentiating neurons
        Jordan Dizon presents webinar on Differentiating neurons
        30:12
      • Play video Heart disease in a dish: Human iPSC-derived multicellular cardiac organoids
        Heart disease in a dish: Human iPSC-derived multicellular cardiac organoids
        This presentation showcases the development of an advanced 3D multicellular human heart tissue model for modelling cardiovascular diseases.
        51:39
      • Play video Perfect Pairings: beta-3 Tubulin Monoclonal Antibody and ProLong RapidSet Antifade Mountant
        Perfect Pairings: beta-3 Tubulin Monoclonal Antibody and ProLong RapidSet Antifade Mountant
        Enable fast, reliable labeling and imaging of neuronal markers.
        0:17
      • Play video Stem cell research-How to use Gibco® PSC Neural Induction Medium
        Stem cell research-How to use Gibco® PSC Neural Induction Medium
        Generate at least 20 million NSCs from 1 million PSCs in just 7 days using Gibco® PSC Neural Induction medium. Learn how to passage and plate PSCs at the proper density and which inhibitors to use.
        3:20
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels