How to Label Technical Replicates in Thermo Fisher Scientific Real-Time PCR Software - Ask TaqMan Episode 1
2:42
Description
Related Videos
For the uninitiated, when we say "technical replicates," we're referring to wells that contain exactly the same amount of the same sample, amplified with the same Assay.