Which Quencher Molecule is Best for TaqMan Probes in Real-Time PCR? -- Ask TaqMan Episode 3
4:09
Description
Related Videos
Applied Biosystems real-time instruments officially support two different quenchers, the first of which is an oldie but goodie called TAMRA dye. TAMRA dye has been around for a long time, and it's still widely available.