Homozygous vs. Heterozygous Samples in qPCR - Ask TaqMan #23
4:36
Description
Related Videos
Did you know that TaqMan assays can also be used for SNP Genotyping? While the assays use the same robust TaqMan chemistry, they contain two probes, and thus analysis is different than when using a Gene Expression assay.