      • Stem Cell Research
      Chemiluminescent western blot detection: Bright and bold detection no matter how scarce your target
      40:38
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Chemiluminescent detection is a widely-used method for detecting target proteins in a western blotting. This detection method is the result of an enzyme reaction of horseradish peroxidase (HRP) or alkaline phosphatase (AP) which produces light, and can be measured by use of x-ray film or a CCD imager. Chemiluminescent substrates allow for various advantages over other methods in detection including, but not limited to, increased sensitivity, high signal-to noise ratios (or low background), and the ability to strip and reprobe the blot. Contrary to popular belief, however, "one size" does not fit all" when it comes to western blotting substrates. With so many options on the market, it is sometimes difficult to determine which substrate performs the best for individual western blot systems. In this webinar we will discuss the best methods for optimizing this technique as well as how to determine the appropriate reagents for your sensitive target detection.
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels