Thermo Scientific Nunc Cell Factory Shaker Overview
1:19
Description
Related Videos
Watch this video to learn more about the Thermo Scientific Nunc Cell Factory Shaker. This cell factory equipment is an automated solution to detach cells from the Thermo Scientific Nunc Cell Factory system. It eliminates the traditional need for manual methods of cell detachment, such as shaking or pounding and complies with the safety requirements, standards, and guidelines.