Thermo Scientific Nunc Cell Factory Incubator overview
2:06
Description
Related Videos
Watch this video to learn more about the Thermo Scientific Nunc Cell Factory Incubator. It is a mobile solution for the incubation of Nunc Cell Factory systems. Mitigate the risk of spillage from moving full Cell Factory systems manually. The convenient floor-level height of the shelves makes it easy to load and unload the Cell Factory carts. This equipment is designed to seamlessly accommodate four racks of 10-layer or 40-layer Nunc Cell Factory systems.