      • Stem Cell Research
      Thermo Scientific Nunc Automatic Cell Factory Manipulator System Overview
      4:29
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Watch this video to learn more about the Thermo Scientific Nunc Automatic Cell Factory Manipulator. This cell factory equipment automates the filling and harvesting steps for twelve 10- or 13-layer and four 40- or 52-layer Cell Factory systems. Using this automatic manipulator, you can increase your processing speed by eliminating manual filling, emptying, harvesting and trypsinization steps of all Cell Factory systems.
      View More
      View Less
      Learn more about Nunc Cell Factory Automation Equipment
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels