      • Stem Cell Research
      Pro-Detect Rapid Assay Lateral Flow Kits for Detecting Protein Tags and Antibody Isotyping
      32:32
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Lateral flow assays (LFAs) are rapid, sensitive, and easy-to-use immunoassays for detecting specific target proteins in vitro. Although the LFA format is extremely popular in point-of-care diagnostic tests (i.e., pregnancy tests), to-date its use in basic research laboratories is not widespread. Here we present the Thermo Scientific Pro-Detect Rapid Assay, an LFA designed for research workflows that can be used for rapid monitoring of expressed recombinant proteins and rapid identification of antibody isotypes. The Pro-Detect Rapid Assay is an alternative to western blotting and ELISA formats that require significantly more time for sample processing as well as additional instrumentation. The rapid qualitative (yes/no) determination of this LFA will also help address key pain points in cell culture and protein expression workflows. Learning Objectives Understand how lateral flow technology–based Pro-Detect Rapid Assays can be used to rapidly detect protein expression Understand how lateral flow technology–based Pro-Detect Rapid Assays can be used to rapidly isotype human, mouse, or rat antibodies
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels