      • Stem Cell Research
      Protein Sample Evaluation Using the Nanodrop One UV-Vis Spectrophotometer
      56:37
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Protein sample evaluation is an important step in many protein workflows. The NanoDrop One microvolume spectrophotometer supports protein sample quantification with multiple preconfigured applications, including direct absorbance at A280, A205, or other wavelengths using the Thermo Scientific Pierce colorimetric assays (BCA, Rapid Gold BCA, Pierce 660, Bradford, etc.) and a proteins and labels application for quantitation of metalloproteins and fluorescently labeled proteins. We will review what you need to consider when choosing a protein quantification method for your sample. Is the sample one purified protein or a mixture of proteins? Does your peptide contain tyrosine and tryptophan residues? Does the sample buffer absorb in the UV spectrum? How can co-purified DNA affect your protein sample measurement? What is a good blank/control for your sample? Answers to these questions and other key considerations will be discussed. Learning objectives: Understand protein extinction coefficients—how to obtain the most accurate protein sample quantification using UV absorbance at 280 nm Learn how new Thermo Scientific Acclaro sample intelligence technology in the NanoDrop One spectrophotometer delivers information not only about the concentration but also the quality of your protein sample
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels