      • Stem Cell Research
      Cells-to-CT Kits: A fast way to kill your cells for gene expression analysis
      32:45
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Real-time reverse transcription PCR analysis of RNA expression in normal and diseased states is a fundamental technique used in research and screening workflows. However, traditional purification of RNA samples often represents a significant workflow bottleneck, which increases the time and automation complexity required for analysis. We describe the Cells-to-CT technology, a workflow that was designed in conjunction with real-time analysis to completely eliminate the need for traditional purification while maintaining performance relative to purified RNA. High-density sample preparation is accomplished in less than 8 min and is highly amenable to automation. We demonstrate that results are equivalent to RNA purified using traditional methods over a broad range of sample input while offering advantages when dealing with automated liquid handlers. Watch this webinar presentation by the developer of Invitrogen™ Cells-to-CT™ kits, Laura Chapman, Staff Scientist in R&D at Thermo Fisher Scientific. You’ll learn how Cells-to-CT kits enable you to measure relative gene expression by real-time qRT-PCR without having to purify the RNA prior to amplification. Cells-to-CT kits are available with Invitrogen™ TaqMan® and SYBR™ Green detection chemistries and enable real-time qRT-PCR analysis directly from cultured cells. Cells-to-CT kits help you to get true value with excellent performance and reliability.
      View More
      View Less
      Find out more about our Cells-to-CT kits
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels