      • Stem Cell Research
      How to strip and reprobe your western blot
      1:55
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Stripping and reprobing a western blot save time and conserves samples while allowing for optimization or detection of a different target protein. With chemiluminescent and fluorescent detection, the signal-producing chemicals can be washed away, and the antibody stripped from the membrane so that the blot can be reprobed for another target protein. If you’re going to be performing multiple strips and reprobing for different antigens, probe for the low-abundance proteins first. Various stripping buffer formulations are commercially available or can be made in the lab. These buffers generally include a detergent and reducing agent. Some formulations require heat and/or low pH to effectively strip off antibodies. To strip a blot of all antibodies and or chemiluminescent reagents, immerse the blot in stripping buffer and either incubate with agitation or heat in an oven. If you are using Thermo Scientific™ Restore™ buffer, incubate the blot for 5 to 10 minutes at 37 degrees Celsius. If using Restore for Fluorescent blots, incubate 10–15 minutes at room temperature. After the specified incubation period, wash the blot thoroughly to ensure that all of the stripping buffer is removed. When using stripping buffers with beta-mercaptoethanol as the reducing agent, you will know that your blot has been sufficiently washed when you can no longer smell the pungent aroma of the reagent. Once stripped, the blot will need to be re-blocked before the second immunoprobing experiment can be performed. Learn more information about western blot stripping buffers at thermofisher.com/western buffers
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels