      • Stem Cell Research
      Process 11 Parallel Twin-Screw Extruder - Overview
      5:34
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Starting with as little as 20 g/h of material, the Thermo Scientific Process 11 Twin-screw Extruder is designed to meet the key challenges faced by research and development formulation scientists. Complete compounding line including feeding solutions and downstream equipment such as water baths, conveyor belt, variable length pelletizers, melt pump, 3D filament spooler or sheet take off are available.
      View More
      View Less
      Learn more about Process 11 Twin-Screw Extruder capabilities and applications
      Share
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels