AAV downstream processing for industrial scale production - moving towards commercial manufacturing
1:01:04
Description
Related Videos
The utilization of POROSâ„¢ CaptureSelectâ„¢ AAV affinity resins results in a significant improvement to the downstream process of AAV. In conjunction, the importance of understanding regulatory requirements such as process and product impurity clearance and viral clearance are discussed and results of a viral clearance study are shown. This study demonstrates that the POROSâ„¢ CaptureSelectâ„¢ AAVX affinity resin could serve as an effective viral clearance removal step in the downstream manufacturing of these vectors.