      • Stem Cell Research
      Three questions to ask when choosing a PCR enzyme
      1:17
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Do you need to preserve sequence accuracy? If not, a standard DNA polymerase can be used to confirm presence or absence of target DNA. If, however, you will clone or perform sequencing, choose a high-fidelity proofreading polymerase for accurate sequence. Are you working with long DNA templates or looking to cut down cycling time? If so, choose a polymerase with a high processivity and extension rate. This will ensure efficient DNA synthesis and shorter reaction times. Are you encountering non-specific amplification? If you’re seeing too many bands that should not be there, choose a hot-start polymerase. This can help get more yield and sensitivity of the target.
      View More
      View Less
      Learn more about choosing PCR enzymes
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels