      • Stem Cell Research
      Boosting lab productivity with PCR enzymes
      1:00
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Are you looking to boost your lab efficiency and productivity? Consider robust PCR enzymes that are designed for speed, simplicity, and accuracy. Speed: You can enjoy short PCR cycling time from fast DNA synthesis of Invitrogen Platinum II Taq Hot-Start DNA Polymerase at 15 sec/kb. Simplicity: You can shorten your PCR workflow by ~50% with Invitrogen Platinum Direct PCR Universal Master Mix. This product requires no DNA purification prior to PCR. Accuracy: You can spend less time troubleshooting downstream applications because amplicons of highly accurate sequences are generated by Invitrogen Platinum SuperFi II DNA Polymerase. This enzyme provides >300x higher fidelity than Taq DNA polymerase does.
      View More
      View Less
      Find your PCR boosters
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels