      • Stem Cell Research
      Six PCR tips for quick mouse genotyping
      1:51
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Here are six PCR tips for quick mouse genotyping. 1. Skip genomic DNA extraction. Use a direct PCR master mix which allows you to add tissue directly to the PCR reaction, without the need for genomic DNA purification. 2. Optimize PCR annealing, faster. Use a direct PCR master mix that enables a universal annealing temperature of 60°C. 3. Handle fewer reagents tubes. Use a master mix format that is premixed with necessary PCR components. Only your sample and primers need to be added. 4. Finish PCR in minutes, not hours. Choose a polymerase that can synthesize DNA at 15–20 sec/kb for fast cycling. 5. Load PCR reactions directly onto gels. Use a direct PCR master mix containing electrophoresis dyes for direct gel loading. Skip addition of the dyes to the reactions for electrophoresis. 6. Complete electrophoresis in 15 min or less. Use bufferless precast gels and an electrophoresis system integrated with a camera hood. Just load, run, and analyze your PCR samples.
      View More
      View Less
      Learn more about the genotyping PCR tips
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels