      • Stem Cell Research
      Collibri 3' mRNA Library Prep for Illumina Systems
      2:56
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Low cost gene expression studies are now possible from as few as 2 million reads in a single-end, 75 bp sequencing run on Illumina NGS systems. The Invitrogen Collibri 3’ mRNA Library Prep Kits for Illumina Systems use a blocking mechanism to capture only the 3’ end of each transcript, enabling shorter sequencing runs. Normalization (RPKM) is not required with this sequencing method because the transcript is degraded during library preparation, ensuring that gene expression results match sample complexity in the sequencing data. Globin-blocking oligos are included in the Invitrogen Collibri 3’ Blood mRNA Kit to enrich for gene expression by reducing the presence of globin in sequencing data. Collibri 3’ mRNA kits are compatible with all samples types and quality, including FFPE. Collibri 3’ Blood mRNA kits are compatible with all human samples.
      View More
      View Less
      Share
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels